Anti-CK19 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512431
CAPIPA5-12431EA 2561.45 CAD
CAPIPA5-12431
Anti-CK19 Rabbit Polyclonal Antibody
KRT19 is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. Unlike its related family members, this smallest known acidic cytokeratin is not paired with a basic cytokeratin in epithelial cells. It is specifically expressed in the periderm, the transiently superficial layer that envelopes the developing epidermis.

Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: CK19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR