Anti-PFDN6 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512450
CAPIPA5-12450EA 2561.45 CAD
CAPIPA5-12450
Anti-PFDN6 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine and mouse based on sequence homology. PFDN6 binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. This protein also binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PFDN6 (prefoldin subunit 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR