Anti-SNRPD1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512460
CAPIPA5-12460EA 2561.45 CAD
CAPIPA5-12460
Anti-SNRPD1 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, mouse and non-human primate based on sequence homology. SNRPD1 is a small nuclear ribonucleoprotein that belongs to the SNRNP core protein family. This protein may act as a charged protein scaffold to promote SNRNP assembly or strengthen SNRNP-SNRNP interactions through nonspecific electrostatic contacts with RNA.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: SNRPD1 (small nuclear ribonucleoprotein D1 polypeptide 16kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR