Anti-ENPP2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512478
CAPIPA5-12478EA 2801.79 CAD
CAPIPA5-12478
Anti-ENPP2 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine based on sequence homology. ENPP2 functions as both a phosphodiesterase, which cleaves phosphodiester bonds at the 5' end of oligonucleotides, and a phospholipase, which catalyzes production of lysophosphatidic acid (LPA) in extracellular fluids. LPA evokes growth factor-like responses including stimulation of cell proliferation and chemotaxis. This protein stimulates the motility of tumor cells and has angiogenic properties, and its expression is upregulated in several kinds of carcinomas. This protein is secreted and further processed to make the biologically active form.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: ENPP2 (ectonucleotide pyrophosphatase/phosphodiesterase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR