Anti-HNRNPK Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512481
CAPIPA5-12481EA 2444.36 CAD
CAPIPA5-12481
Anti-HNRNPK Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, mouse, non-human primate, rat and rabbit based on sequence homology. HNRPK belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. HNRPK is located in the nucleoplasm and has three repeats of KH domains that binds to RNAs. The protein is distinct among other hnRNP proteins in its binding preference; it binds tenaciously to poly(C). This protein is also thought to have a role during cell cycle progession.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HNRNPK (heterogeneous nuclear ribonucleoprotein K)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR