Anti-HSPH1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512485
CAPIPA5-12485EA 2561.45 CAD
CAPIPA5-12485
Anti-HSPH1 Rabbit Polyclonal Antibody
HSPH1 prevents the aggregation of denatured proteins in cells under severe stress, on which the ATP levels decrease markedly. This protein inhibits HSPA8/HSC70 ATPase and chaperone activities.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HSPH1 (heat shock 105kDa/110kDa protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR