Anti-RFC3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512486
CAPIPA5-12486EA 2561.45 CAD
CAPIPA5-12486
Anti-RFC3 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine and mouse based on sequence homology. The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kDa.

Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: RFC3 (replication factor C (activator 1) 3, 38kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR