Anti-AIMP2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512487
CAPIPA5-12487EA 2561.45 CAD
CAPIPA5-12487
Anti-AIMP2 Rabbit Polyclonal Antibody
JTV1 is probable core protein of the multisynthetase complex that serves as a template for the assembly of the supramolecular structure. This protein Mediates ubiquitination of FUBP1 and its degradation by the proteasome.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: AIMP2 (aminoacyl tRNA synthetase complex-interacting multifunctional protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR