Anti-SLAM1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512492
CAPIPA5-12492EA 2801.79 CAD
CAPIPA5-12492
Anti-SLAM1 Rabbit Polyclonal Antibody
Antibodies
SLAMF1 is high-affinity self-ligand important in bidirectional T-cell to B-cell stimulation. SLAM-induced signal-transduction events in T-lymphocytes are different from those in B-cells. Two modes of SLAM signaling are likely to exist: one in which the inhibitor SH2D1A acts as a negative regulator and another in which protein-tyrosine phosphatase 2C (PTPN11)-dependent signal transduction operates.

Western Blot: 1:1000

Type: Primary
Antigen: SLAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR