Anti-CDA Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512497
CAPIPA5-12497EA 2561.45 CAD
CAPIPA5-12497
Anti-CDA Rabbit Polyclonal Antibody
CDA forms a homotetramer that catalyzes the irreversible hydrolytic deamination of cytidine and deoxycytidine to uridine and deoxyuridine, respectively. This protein is one of several deaminases responsible for maintaining the cellular pyrimidine pool. Mutations in the CDA gene are associated with decreased sensitivity to the cytosine nucleoside analogue cytosine arabinoside used in the treatment of certain childhood leukemias.

Western Blot: 1:1000

Type: Primary
Antigen: CDA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR