Anti-CD38 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512509
CAPIPA5-12509EA 2801.79 CAD
CAPIPA5-12509
Anti-CD38 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with non-human primate based on sequence homology. CD38 is a novel multifunctional ectoenzyme widely expressed in cells and tissues especially in leukocytes. CD38 also functions in cell adhesion,signal transduction and calcium signaling.

Flow Cytometry: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunofluorescence: 1:10-1:50, Immunohistochemistry (Paraffin): 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: CD38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR