Anti-ENG Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512511
CAPIPA5-12511EA 2801.79 CAD
CAPIPA5-12511
Anti-ENG Rabbit Polyclonal Antibody
Antibodies
CD105 is a homodimeric transmembrane protein which is a major glycoprotein of the vascular endothelium. This protein is a component of the transforming growth factor beta receptor complex and it binds TGFB1 and TGFB3 with high affinity. Mutations in its gene cause hereditary hemorrhagic telangiectasia, also known as Osler-Rendu-Weber syndrome 1, an autosomal dominant multisystemic vascular dysplasia.

Flow Cytometry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: ENG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR