Anti-ELP2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512515
CAPIPA5-12515EA 2561.45 CAD
CAPIPA5-12515
Anti-ELP2 Rabbit Polyclonal Antibody
ELP2 regulates the ligand-dependent activation of STAT3. The protein acts as subunit of the RNA polymerase II elongator complex, which is a histone acetyltransferase component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:100, Immunocytochemistry: 1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: ELP2 (elongator acetyltransferase complex subunit 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR