Anti-CAPZB Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512521
CAPIPA5-12521EA 2561.45 CAD
CAPIPA5-12521
Anti-CAPZB Rabbit Polyclonal Antibody
CAPZB is a member of the F-actin capping protein family. This protein is the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.

Western Blot: 1:1000

Type: Primary
Antigen: CAPZB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR