Anti-HIST3H3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512526
CAPIPA5-12526EA 2831.39 CAD
CAPIPA5-12526
Anti-HIST3H3 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, chicken, canine, equine, mouse, porcine, rat, rabbit and Xenopus, yeast and zebrafish based on sequence homology. Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. This structure consists of approximately 146 bp of DNA wrapped around a nucleosome, an octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. The gene for this protein is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. The gene is found in the large histone gene cluster on chromosome 6p22-p21.3.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR