Anti-ABL2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512537
CAPIPA5-12537EA 2561.45 CAD
CAPIPA5-12537
Anti-ABL2 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. ABL2 is a cytoplasmic tyrosine kinase which is closely related to but distinct from ABL1. The similarity of the proteins includes the tyrosine kinase domains and extends amino-terminal to include the SH2 and SH3 domains. ABL2 is expressed in both normal and tumor cells.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Dot blot: 1:500

Type: Primary
Antigen: ABL2 (c-abl oncogene 2, non-receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR