Anti-c-Abl Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512538
CAPIPA5-12538EA 2561.45 CAD
CAPIPA5-12538
Anti-c-Abl Rabbit Polyclonal Antibody
ABL2 is a cytoplasmic tyrosine kinase which is closely related to but distinct from ABL1. The similarity of the proteins includes the tyrosine kinase domains and extends amino-terminal to include the SH2 and SH3 domains. ABL2 is expressed in both normal and tumor cells. The ABL2 gene product is expressed as two variants bearing different amino termini, both approximately 12-kb in length. The peptide used to generate this antibody is 100% conserved between ABL2 and ABL1.

Western Blot: 1:1000

Type: Primary
Antigen: c-Abl
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR