Anti-CDK2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512572
CAPIPA5-12572EA 2573.22 CAD
CAPIPA5-12572
Anti-CDK2 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, hamster, mouse and rat based on sequence homology. Cdk2 is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This protein forms a trimeric complex with cyclin H and MAT1, which functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This protein is thought to serve as a direct link between the regulation of transcription and the cell cycle.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CDK2 (cyclin-dependent kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR