Anti-HIST1H3B3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512604
CAPIPA5-12604EA 2561.45 CAD
CAPIPA5-12604
Anti-HIST1H3B3 Rabbit Polyclonal Antibody
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. This structure consists of approximately 146 bp of DNA wrapped around a nucleosome, an octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6p22-p21.3.

Recommended Dilutions: Western Blot: 1:2000, Immunohistochemistry: 1:50-1:100, Dot blot: 1:500

Type: Primary
Antigen: HIST1H3B3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR