Anti-ERBB4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512607
CAPIPA5-12607EA 2573.22 CAD
CAPIPA5-12607
Anti-ERBB4 Rabbit Polyclonal Antibody
Antibodies
The HER4/ERBB4 gene is a member of the type I receptor tyrosine kinase subfamily that includes EGFR, ERBB2, and ERBB3. It encodes a receptor for NDF/heregulin.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: ERBB4 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR