Anti-LPR1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512617
CAPIPA5-12617EA 2128.34 CAD
CAPIPA5-12617
Anti-LPR1 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. LPR1 is involved in the plasma clearance of chylomicron remnants and activated alpha 2-macroglobulin, as well as the local metabolism of complexes between plasminogen activators and their endogenous inhibitors.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100, Dot blot: 1:500

Type: Primary
Antigen: LPR1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR