Anti-MAPKAPK5 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512621
CAPIPA5-12621EA 2286.72 CAD
CAPIPA5-12621
Anti-MAPKAPK5 Rabbit Polyclonal Antibody
Antibodies
The protein encoded by this gene is a member of the serine/threonine kinase family. In response to cellular stress and proinflammatory cytokines, this kinase is activated through its phosphorylation by MAP kinases including MAPK1/ERK, MAPK14/p38-alpha, and MAPK11/p38-beta. In vitro, this kinase phosphorylates heat shock protein HSP27 at its physiologically relevant sites.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: MAPKAPK5 (mitogen-activated protein kinase-activated protein kinase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR