Anti-MYT1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512633
CAPIPA5-12633EA 2801.79 CAD
CAPIPA5-12633
Anti-MYT1 Rabbit Polyclonal Antibody
Antibodies
The protein encoded by this gene is a member of the serine/threonine protein kinase family. This kinase preferentially phosphorylates and inactivates cell division cycle 2 protein (CDC2), and thus negatively regulates cell cycle G2/M transition. This kinase is associated with the membrane throughout the cell cycle. Its activity is highly regulated during the cell cycle. Protein kinases AKT1/PKB and PLK (Polo-like kinase) have been shown to phosphorylate and regulate the activity of this kinase. Alternatively spliced transcript variants encoding distinct isoforms have been reported.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Dot blot: 1:500

Type: Primary
Antigen: MYT1 (myelin transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR