Anti-RB1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512679
CAPIPA5-12679EA 2573.22 CAD
CAPIPA5-12679
Anti-RB1 Rabbit Polyclonal Antibody
Antibodies
Retinoblastoma (RB) is an embryonic malignant neoplasm of retinal origin. It almost always presents in early childhood and is often bilateral. Spontaneous regression ('cure') occurs in some cases.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100, Dot blot: 1:500

Type: Primary
Antigen: RB1 (retinoblastoma 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR