Anti-SEPARIN Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512691
CAPIPA5-12691EA 2573.22 CAD
CAPIPA5-12691
Anti-SEPARIN Rabbit Polyclonal Antibody
Antibodies
The metaphase-to-anaphase transition is the final discrete event in duplication and separation of the genetic material of the cell. Its timing is regulated by the activation of the anaphase-promoting complex (APC). In both budding and fission yeast, the degradation of the Pds1 or Cut2 protein, respectively, is required for the onset of sister chromatid separation. Both proteins are APC substrates. Pds1 and Cut2 proteins associate with the yeast separin proteins Esp1 and Cut1, respectively, and prevent the separins from promoting chromatid separation. Pds1 and Cut2 are also called anaphase inhibitors or securins

Recommended Dilutions: Immunohistochemistry: 1:50-1:100, Dot blot: 1:500

Type: Primary
Antigen: SEPARIN
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR