Anti-SMAD4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512695
CAPIPA5-12695EA 2801.79 CAD
CAPIPA5-12695
Anti-SMAD4 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. Common mediator of signal transduction by TGF-beta (transforming growth factor) superfamily, SMAD4 is the common SMAD (co-SMAD). It promotes binding of the SMAD2/SMAD4/FAST-1 complex to DNA and provides an activation function required for SMAD1 or SMAD2 to stimulate transcription. It may act as a tumor suppressor.

Recommended Dilutions: Immunofluorescence: 1:10-1:50, Immunohistochemistry: 1:10-1:50, Dot blot: 1:500

Type: Primary
Antigen: SMAD4 (SMAD family member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR