Anti-TNIK Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512711
CAPIPA5-12711EA 2128.34 CAD
CAPIPA5-12711
Anti-TNIK Rabbit Polyclonal Antibody
TNIK is a stress-activated serine/threonine kinase that may play a role in the response to environmental stress. This protein appears to act upstream of the JUN N-terminal pathway, and may play a role in cytoskeletal regulation.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100, Dot blot: 1:500

Type: Primary
Antigen: TNIK (TRAF2 and NCK interacting kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR