Anti-S6K Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512723
CAPIPA5-12723EA 2561.45 CAD
CAPIPA5-12723
Anti-S6K Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, mouse, rat and rabbit based on sequence homology. RPS6KB1 is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates several residues of the S6 ribosomal protein. The kinase activity of this protein leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding the gene for RPS6KB1 and overexpression of this kinase are seen in some breast cancer cell lines.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: S6K
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR