Anti-JUN Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512733
CAPIPA5-12733EA 2572.49 CAD
CAPIPA5-12733
Anti-JUN Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, chicken, mouse, porcine and rat based on sequence homology. This gene for the cJun protein is the putative transforming gene of avian sarcoma virus 17. The protein is highly similar to the viral protein, and interacts directly with specific target DNA sequences to regulate gene expression. The gene for this protein is intronless and is mapped to 1p32-p31, a chromosomal region involved in both translocations and deletions in human malignancies.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: JUN (jun proto-oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR