Anti-RAF1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512765
CAPIPA5-12765EA 2831.39 CAD
CAPIPA5-12765
Anti-RAF1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. Raf-1 is a MAP kinase kinase kinase (MAP3K) which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated Raf-1 can phosphorylate to activate the dual specificity protein kinases MEK1 and MEK2 which in turn phosphorylate to activate the serine/threonine specific protein kinases ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: RAF1 (v-raf-1 murine leukemia viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR