Anti-NR4A1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512771
CAPIPA5-12771EA 2801.79 CAD
CAPIPA5-12771
Anti-NR4A1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. NR4A1 is a member of the steroid-thyroid hormone-retinoid receptor superfamily. Expression is induced by phytohemagglutinin in human lymphocytes and by serum stimulation of arrested fibroblasts. The encoded protein acts as a nuclear transcription factor. Translocation of the protein from the nucleus to mitochondria induces apoptosis.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: NR4A1 (nuclear receptor subfamily 4, group A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR