Anti-ATF2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512802
CAPIPA5-12802EA 2572.49 CAD
CAPIPA5-12802
Anti-ATF2 Rabbit Polyclonal Antibody
Antibodies
ATF2 is a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds to the cAMP-responsive element (CRE), an octameric palindrome. The protein forms a homodimer or heterodimer with c-Jun and stimulates CRE-dependent transcription. The protein is also a histone acetyltransferase (HAT) that specifically acetylates histones H2B and H4 in vitro; thus it may represent a class of sequence-specific factors that activate transcription by direct effects on chromatin components.

  • Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: ATF2 (activating transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR