Anti-BRAF Rabbit Polyclonal Antibody

PA512803 PA512803
CAPIPA5-12803EA 1227.26 CAD
PIPA5-12803 CAPIPA5-12803
Anti-BRAF Rabbit Polyclonal Antibody
This antibody is predicted to react with chicken and mouse based on sequence homology. BRAF is involved in the transduction of mitogenic signals from the cell membrane to the nucleus. It may play a role in the postsynaptic responses of hippocampal neuron. Defects in BRAF are a cause of cardiofaciocutaneous syndrome (CFC syndrome), and a wide range of cancers such as lung cancer, non-Hodgkins lymphoma, and colorectal cancer.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: BRAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR