Anti-MEF2C Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512811
CAPIPA5-12811EA 2573.22 CAD
CAPIPA5-12811
Anti-MEF2C Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, mouse and porcine based on sequence homology. MEF2C is a transcription activator which binds specifically to the MEF2 element present in the regulatory regions of many muscle-specific genes. This protein controls cardiac morphogenesis and myogenesis, and is also involved in vascular development. It may also be involved in neurogenesis and in the development of cortical architecture.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR