Anti-NFATC2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512859
CAPIPA5-12859EA 2286.72 CAD
CAPIPA5-12859
Anti-NFATC2 Rabbit Polyclonal Antibody
Antibodies
NFATC2 is a member of the nuclear factor of activated T cells (NFAT) family. It is a DNA-binding protein with a REL-homology region (RHR) and an NFAT-homology region (NHR). This protein is present in the cytosol and only translocates to the nucleus upon T cell receptor (TCR) stimulation, where it becomes a member of the nuclear factors of activated T cells transcription complex. This complex plays a central role in inducing gene transcription during the immune response.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: NFATC2 (nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR