Anti-RPS6KA3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512892
CAPIPA5-12892EA 2561.45 CAD
CAPIPA5-12892
Anti-RPS6KA3 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse and zebrafish based on sequence homology. RSK3 is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in the gene have been associated with Coffin-Lowry syndrome (CLS).

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: RPS6KA3 (ribosomal protein S6 kinase, 90kDa, polypeptide 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR