Anti-PIK3CD Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512922
CAPIPA5-12922EA 2572.49 CAD
CAPIPA5-12922
Anti-PIK3CD Rabbit Polyclonal Antibody
Antibodies
Phosphoinositide 3-kinases (PI3Ks) phosphorylate the 3-prime OH position of the inositol ring of inositol lipids. See MIM 602838. The class I PI3Ks display a broad phosphoinositide lipid substrate specificity and include p110-alpha, p110-beta, and p110-gamma. p110-alpha and p110-beta interact with SH2/SH3-domain-containing p85 adaptor proteins and with GTP-bound Ras.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: PIK3CD (phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR