Anti-EGFR Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512936
CAPIPA5-12936EA 2831.39 CAD
CAPIPA5-12936
Anti-EGFR Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse based on sequence homology. Epidermal Growth factor receptor (EGFR) is the prototype member of the type 1 receptor tyrosine kinases. EGFR overexpression in tumors indicates poor prognosis and is observed in tumors of the head and neck, brain, bladder, stomach, breast, lung, endometrium, cervix, vulva, ovary, esophagus, stomach and in squamous cell carcinoma. EGFR is a receptor for EGF, but also for other members of the EGF family, including TGF-alpha, amphiregulin, betacellulin, heparin-binding EGF-like growth factor, GP30 and vaccinia virus growth factor. It is involved in the control of cell growth and differentiation.

Recommended Dilutions: Western Blot: 1:8000, Dot blot: 1:500

Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR