Anti-CDC25A Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512951
CAPIPA5-12951EA 2713.62 CAD
CAPIPA5-12951
Anti-CDC25A Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. CDC25A is a member of the CDC25 family of phosphatases.CDC25A is required for progression from G1 to the S phase of the cell cycle. It activates the cyclin-dependent kinase CDC2 by removing two phosphate groups. CDC25A is specifically degraded in response to DNA damage, which prevents cells with chromosomal abnormalities from progressing through cell division.CDC25A is an oncogene, although its exact role in oncogenesis has not been demonstrated.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR