Anti-DNMT1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512963
CAPIPA5-12963EA 2573.22 CAD
CAPIPA5-12963
Anti-DNMT1 Rabbit Polyclonal Antibody
Antibodies
DNA (cytosine-5-)-methyltransferase 1 has a role in the establishment and regulation of tissue-specific patterns of methylated cytosine residues. Aberrant methylation patterns are associated with certain human tumors and developmental abnormalities.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: DNMT1 (DNA (cytosine-5-)-methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR