Anti-SLAMF1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512970
CAPIPA5-12970EA 2573.22 CAD
CAPIPA5-12970
Anti-SLAMF1 Rabbit Polyclonal Antibody
Antibodies
SLAMF1 is a high-affinity self-ligand important in bidirectional T-cell to B-cell stimulation. SLAM-induced signal-transduction events in T-lymphocytes are different from those in B-cells. Two modes of SLAM signaling are likely to exist: one in which the inhibitor SH2D1A acts as a negative regulator and another in which protein-tyrosine phosphatase 2C (PTPN11)-dependent signal transduction operates.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: SLAMF1 (signaling lymphocytic activation molecule family member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR