Anti-BAR2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512977
CAPIPA5-12977EA 2353.26 CAD
CAPIPA5-12977
Anti-BAR2 Rabbit Polyclonal Antibody
Antibodies
Beta-2-adrenergic receptor is a member of the G protein-coupled receptor superfamily. This receptor is directly associated with one of its ultimate effectors, the class C L-type calcium channel Ca(V)1.2. This receptor-channel complex also contains a G protein, an adenylyl cyclase, cAMP-dependent kinase, and the counterbalancing phosphatase, PP2A. The assembly of the signaling complex provides a mechanism that ensures specific and rapid signaling by this G protein-coupled receptor.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: BAR2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR