Anti-GIT1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512982
CAPIPA5-12982EA 2286.08 CAD
CAPIPA5-12982
Anti-GIT1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. GIT1 is a GTPase-activating protein for the ADP ribosylation factor family. It may serve as a scaffold to bring together molecules to form signaling modules controlling vesicle trafficking, adhesion and cytoskeletal organization, increases the speed of cell migration, as well as the size and rate of formation of protrusions, possibly by targeting PAK1 to adhesions and the leading edge of lamellipodia. It sequesters inactive non-tyrosine-phosphorylated paxillin in cytoplasmic complexes.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: GIT1 (G protein-coupled receptor kinase interacting ArfGAP 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR