Anti-GFAP Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512991
CAPIPA5-12991EA 2801.79 CAD
CAPIPA5-12991
Anti-GFAP Rabbit Polyclonal Antibody
Antibodies
GFAP is one of the major intermediate filament proteins of mature astrocytes. It is used as a marker to distinguish astrocytes from other glial cells during development. Mutations in this gene cause Alexander disease, a rare disorder of astrocytes in the central nervous system.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR