Anti-TEK Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512992
CAPIPA5-12992EA 2286.72 CAD
CAPIPA5-12992
Anti-TEK Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, mouse and zebrafish based on sequence homology. The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells in mice, rats, and humans. This receptor possesses a unique extracellular domain containing 2 immunoglobulin-like loops separated by 3 epidermal growth factor-like repeats that are connected to 3 fibronectin type III-like repeats. The ligand for the receptor is angiopoietin-1. Defects in TEK are associated with inherited venous malformations; the TEK signaling pathway appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis. TEK is closely related to the TIE receptor tyrosine kinase.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: TEK (TEK tyrosine kinase, endothelial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR