Anti-NOMO1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512994
CAPIPA5-12994EA 2573.22 CAD
CAPIPA5-12994
Anti-NOMO1 Rabbit Polyclonal Antibody
Antibodies
NOMO1 was originally thought to be related to the collagenase family. The gene encoding NOMO1 is one of three highly similar genes in a region of duplication located on the p arm of chromosome 16. These three genes encode closely related proteins that may have the same function. The protein encoded by one of these genes has been identified as part of a protein complex that participates in the Nodal signaling pathway during vertebrate development.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: NOMO1 (NODAL modulator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR