Anti-PRL Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513002
CAPIPA5-13002EA 2573.22 CAD
CAPIPA5-13002
Anti-PRL Rabbit Polyclonal Antibody
Antibodies
Prolactin (PRL) acts primarily on the mammary gland by promoting lactation.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: PRL (prolactin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR