Anti-ZBTB16 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513004
CAPIPA5-13004EA 2883.24 CAD
CAPIPA5-13004
Anti-ZBTB16 Rabbit Polyclonal Antibody
Antibodies
ZBTB16 is a member of the Krueppel C2H2-type zinc-finger protein family. It is a zinc finger transcription factor that contains nine Kruppel-type zinc finger domains at the carboxyl terminus. This protein is located in the nucleus, is involved in cell cycle progression, and interacts with a histone deacetylase.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: ZBTB16 (zinc finger and BTB domain containing 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR