Anti-OSR1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513005
CAPIPA5-13005EA 2573.22 CAD
CAPIPA5-13005
Anti-OSR1 Rabbit Polyclonal Antibody
Antibodies
Odd-skipped related I (Odd1) is a zinc finger protein homologous to the Drosophila Odd-skipped class transcription factors that play critical roles in embryonic patterning and tissue morphogenesis.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: OSR1 (odd-skipped related transciption factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR