Anti-LEO1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513010
CAPIPA5-13010EA 2573.22 CAD
CAPIPA5-13010
Anti-LEO1 Rabbit Polyclonal Antibody
Antibodies
The PAF1 complex is a multifunctional complex. The PAF1 complex interacts with POLR2A. May be involved in both initiation and elongation, histone methylation and RNA processing. Overexpression of LEO1 induces cell growth arrest and premature senescence of fibroblasts.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: LEO1 (Leo1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR